Assay Details
Target Gene Details
Entrez Gene ID: | 5786 |
Gene Name: | protein tyrosine phosphatase, receptor type A |
Gene Aliases: |
HEPTP, HLPR, HPTPA, HPTPalpha, LRP, PTPA, PTPRL2, R-PTP-alpha, RPTPA |
Location: |
Chr.20:2864195-3038669 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PTPRA | NM_002836.3 | NP_002827.1 | ||
M34668.1 | AAA36528.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3645020 | Chr.20:2816405 - 2926274 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv1057464 | Chr.20:2650623 - 2989686 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv522009 | Chr.20:2745032 - 3030542 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv833895 | Chr.20:2730164 - 2882236 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
esv2762056 | Chr.20:2740745 - 2944353 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
