Assay Details
Target Gene Details
Entrez Gene ID: | 80025 |
Gene Name: | pantothenate kinase 2 |
Gene Aliases: |
C20orf48, HARP, HSS, NBIA1, PKAN |
Location: |
Chr.20:3888839-3929882 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PANK2 | NM_001324191.1 | 8 | 6947 | NP_001311120.1 |
| NM_001324193.1 | 7 | 5972 | NP_001311122.1 | |
| NM_024960.5 | 7 | 6050 | NP_079236.3 | |
| NM_153638.3 | 7 | 6779 | NP_705902.2 | |
| NM_153640.3 | 7 | 6115 | NP_705904.1 | |
| NR_136715.1 | 7 | 6636 |
Target Gene Details
Entrez Gene ID: | 11237 |
Gene Name: | ring finger protein 24 |
Gene Aliases: |
G1L |
Location: |
Chr.20:3927309-4015591 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF24 | NM_001134337.2 | 6 | 6479 | NP_001127809.1 |
| NM_001134338.2 | 7 | 6548 | NP_001127810.1 | |
| NM_001321749.1 | 6 | 6715 | NP_001308678.1 | |
| NM_007219.4 | 6 | 6771 | NP_009150.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645053 | Chr.20:3849918 - 3987827 on Build GRCh38 | Loss |
|
| nsv828673 | Chr.20:3738382 - 4184811 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs201982152] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map