Assay Details
Target Gene Details
Entrez Gene ID: | 6185 |
Gene Name: | ribophorin II |
Gene Aliases: |
RIBIIR, RPN-II, RPNII, SWP1 |
Location: |
Chr.20:37179046-37241623 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPN2 | NM_001135771.2 | NP_001129243.1 | ||
| NM_001324299.1 | NP_001311228.1 | |||
| NM_001324301.1 | NP_001311230.1 | |||
| NM_001324302.1 | NP_001311231.1 | |||
| NM_001324303.1 | NP_001311232.1 | |||
| NM_001324304.1 | NP_001311233.1 | |||
| NM_001324305.1 | NP_001311234.1 | |||
| NM_001324306.1 | NP_001311235.1 | |||
| NM_002951.4 | NP_002942.2 | |||
| XM_006723851.2 | XP_006723914.1 | |||
| XM_006723852.2 | XP_006723915.1 | |||
| AK096243.1 | ||||
| AK308683.1 | ||||
| AK315801.1 | ||||
| BC003560.1 | AAH03560.1 | |||
| BC020222.1 | AAH20222.1 | |||
| BG718075.1 | ||||
| CR456899.1 | CAG33180.1 | |||
| DB449190.1 | ||||
| Y00282.1 | CAA68393.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3645750 | Chr.20:37049310 - 37196647 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map