Assay Details
Target Gene Details
Entrez Gene ID: | 140836 |
Gene Name: | barrier to autointegration factor 2 |
Gene Aliases: |
BAF-L, BAF2, BAFL, C20orf179 |
Location: |
Chr.20:17693675-17735872 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BANF2 | NM_001159495.1 | NP_001152967.1 | ||
| AW022487.1 |
More Information
Set Membership: |
|

Genomic Map