Assay Details
Target Gene Details
Entrez Gene ID: | 100528031 |
Gene Name: | FKBP1A-SDCBP2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.20:1309909-1393172 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FKBP1A-SDCBP2 | NR_037661.1 | |||
AK309026.1 | ||||
BM925547.1 |
Target Gene Details
Entrez Gene ID: | 100507495 |
Gene Name: | SDCBP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:1325343-1378735 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SDCBP2-AS1 | NR_040047.1 | |||
NR_040048.1 | ||||
NR_040049.1 | ||||
AK055993.1 | ||||
DA094549.1 | ||||
DA274215.1 | ||||
DB454162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3644976 | Chr.20:1327053 - 1337420 on Build GRCh38 | Loss |
![]() ![]() ![]() |
esv2664030 | Chr.20:1327076 - 1337188 on Build GRCh38 | Deletion |
![]() ![]() ![]() |
nsv1057097 | Chr.20:1050297 - 1333621 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
