Assay Details
Target Gene Details
Entrez Gene ID: | 2139 |
Gene Name: | EYA transcriptional coactivator and phosphatase 2 |
Gene Aliases: |
EAB1 |
Location: |
Chr.20:46894624-47188844 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EYA2 | NM_005244.4 | NP_005235.3 | ||
NM_172110.3 | NP_742108.2 | |||
XM_005260327.2 | XP_005260384.1 | |||
XM_017027721.1 | XP_016883210.1 | |||
AK293032.1 | ||||
BC008803.2 | AAH08803.1 | |||
BC013882.2 | AAH13882.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv510788 | Chr.20:46884565 - 46990248 on Build GRCh38 | Deletion |
![]() |
nsv1061899 | Chr.20:46498425 - 47439490 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
