Assay Details
Target Gene Details
Entrez Gene ID: | 140870 |
Gene Name: | WAP four-disulfide core domain 6 |
Gene Aliases: |
C20orf171, HEL-S-295, WAP6, dJ461P17.11 |
Location: |
Chr.20:45534197-45539495 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFDC6 | NM_080827.1 | NP_543017.1 | ||
| AF411861.1 | AAN03684.1 | |||
| BC101600.1 | AAI01601.1 | |||
| BC101604.1 | AAI01605.1 | |||
| GQ472217.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map