Assay Details
Target Gene Details
Entrez Gene ID: | 116154 |
Gene Name: | phosphatase and actin regulator 3 |
Gene Aliases: |
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ |
Location: |
Chr.20:59577497-59849829 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHACTR3 | NM_001199505.1 | NP_001186434.1 | ||
| XM_011528525.2 | XP_011526827.1 | |||
| XM_017027626.1 | XP_016883115.1 | |||
| XM_017027628.1 | XP_016883117.1 | |||
| XM_017027630.1 | XP_016883119.1 | |||
| AK098788.1 |
More Information
Set Membership: |
|

Genomic Map