Assay Details
Target Gene Details
Entrez Gene ID: | 149773 |
Gene Name: | APCDD1L antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.20:58515379-58619892 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APCDD1L-AS1 | NR_034147.1 | |||
| AK054637.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1063462 | Chr.20:58511618 - 58790600 on Build GRCh38 | Gain |
|
| nsv834019 | Chr.20:58397598 - 58607087 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77425254,rs79048259] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map