Assay Details
Target Gene Details
Entrez Gene ID: | 55317 |
Gene Name: | adaptor related protein complex 5 sigma 1 subunit |
Gene Aliases: |
C20orf29 |
Location: |
Chr.20:3820524-3825307 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP5S1 | NM_001204446.1 | NP_001191375.1 | ||
| NM_001204447.1 | NP_001191376.1 | |||
| NM_018347.2 | NP_060817.1 | |||
| AK002030.1 | BAA92045.1 | |||
| AK093332.1 | ||||
| BC043344.1 | AAH43344.1 | |||
| DA129391.1 | ||||
| DA253969.1 | ||||
| DB190907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833899 | Chr.20:3709295 - 3878350 on Build GRCh38 | Loss |
|
| nsv828673 | Chr.20:3738382 - 4184811 on Build GRCh38 | Gain |
|
| esv3645050 | Chr.20:3814525 - 3827525 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map