Assay Details
Target Gene Details
Entrez Gene ID: | 10816 |
Gene Name: | serine peptidase inhibitor, Kunitz type 3 |
Gene Aliases: |
HKIB9 |
Location: |
Chr.20:45512461-45515624 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPINT3 | NM_006652.1 | NP_006643.1 | ||
AY372172.2 | ||||
BX088813.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1056102 | Chr.20:44673341 - 45636248 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
