Assay Details
Target Gene Details
Entrez Gene ID: | 26051 |
Gene Name: | protein phosphatase 1 regulatory subunit 16B |
Gene Aliases: |
ANKRD4, TIMAP |
Location: |
Chr.20:38805693-38923024 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R16B | NM_001172735.2 | NP_001166206.1 | ||
| NM_015568.3 | NP_056383.1 | |||
| XM_017027785.1 | XP_016883274.1 | |||
| AB020630.2 | ||||
| AK311244.1 | ||||
| BC131801.1 | ||||
| BC152467.1 | ||||
| HY125679.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058457 | Chr.20:38277492 - 38844429 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map