Assay Details
Target Gene Details
Entrez Gene ID: | 5203 |
Gene Name: | prefoldin subunit 4 |
Gene Aliases: |
C1, PFD4 |
Location: |
Chr.20:54207963-54219953 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PFDN4 | NM_002623.3 | NP_002614.2 | ||
| XM_017027879.1 | XP_016883368.1 | |||
| AK312059.1 | ||||
| BC010953.1 | AAH10953.1 | |||
| CN482830.1 | ||||
| U41816.1 | AAB17063.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv586224 | Chr.20:53858311 - 54662951 on Build GRCh38 | Gain |
|
| nsv834010 | Chr.20:54143285 - 54316400 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs137871814] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map