Assay Details
Target Gene Details
Entrez Gene ID: | 128553 |
Gene Name: | teashirt zinc finger homeobox 2 |
Gene Aliases: |
C20orf17, OVC10-2, TSH2, ZABC2, ZNF218 |
Location: |
Chr.20:52972309-53495330 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TSHZ2 | NM_173485.5 | NP_775756.3 | ||
XM_017027640.1 | XP_016883129.1 | |||
XM_017027641.1 | XP_016883130.1 | |||
AK091206.1 | BAC03610.1 | |||
BX640770.1 | CAE45871.1 | |||
DA647113.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
