Assay Details
Target Gene Details
Entrez Gene ID: | 101929329 |
Gene Name: | LAMP5 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:9505180-9514998 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LAMP5-AS1 | NR_109957.1 | 5 | 1461 | |
AK096264.1 | 5 | 1461 |
