Assay Details
Target Gene Details
Entrez Gene ID: | 994 |
Gene Name: | cell division cycle 25B |
Gene Aliases: |
- |
Location: |
Chr.20:3786772-3806121 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC25B | NM_001287516.1 | NP_001274445.1 | ||
| NM_001287517.1 | NP_001274446.1 | |||
| NM_001287518.1 | NP_001274447.1 | |||
| NR_136336.1 | ||||
| AK295573.1 | ||||
| AK299028.1 | ||||
| AK299192.1 | ||||
| DB477384.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833899 | Chr.20:3709295 - 3878350 on Build GRCh38 | Loss |
|
| nsv828673 | Chr.20:3738382 - 4184811 on Build GRCh38 | Gain |
|
| nsv585286 | Chr.20:3747323 - 3803946 on Build GRCh38 | Loss |
|
| nsv585285 | Chr.20:3705789 - 3791211 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map