Assay Details
Target Gene Details
Entrez Gene ID: | 57091 |
Gene Name: | Cas scaffolding protein family member 4 |
Gene Aliases: |
C20orf32, CAS4, HEFL, HEPL |
Location: |
Chr.20:56411550-56460240 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASS4 | NM_001164114.1 | NP_001157586.1 | ||
| NM_001164115.1 | NP_001157587.1 | |||
| NM_001164116.1 | NP_001157588.1 | |||
| NM_020356.3 | NP_065089.2 | |||
| XM_006723831.3 | XP_006723894.1 | |||
| AJ276678.1 | CAC00655.1 | |||
| AK027760.1 | BAB55351.1 | |||
| AK295613.1 | ||||
| AK308057.1 | ||||
| AK314653.1 | ||||
| BC027951.1 | AAH27951.1 | |||
| BX280195.1 | ||||
| DA584687.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525425 | Chr.20:56403979 - 56897294 on Build GRCh38 | Gain |
|
| nsv834013 | Chr.20:56307279 - 56476126 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map