Assay Details
Target Gene Details
Entrez Gene ID: | 551 |
Gene Name: | arginine vasopressin |
Gene Aliases: |
ADH, ARVP, AVP-NPII, AVRP, VP |
Location: |
Chr.20:3082555-3093521 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AVP | XM_011529267.1 | 2 | 362 | XP_011527569.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv585276 | Chr.20:3030542 - 3366307 on Build GRCh38 | Gain |
|
| nsv833897 | Chr.20:2990324 - 3175146 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map