Assay Details
Target Gene Details
Entrez Gene ID: | 24141 |
Gene Name: | lysosomal associated membrane protein family member 5 |
Gene Aliases: |
BAD-LAMP, BADLAMP, C20orf103, LAMP-5, UNC-46 |
Location: |
Chr.20:9514358-9530524 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LAMP5 | NM_001199897.1 | NP_001186826.1 | ||
NM_012261.3 | NP_036393.1 | |||
AK295336.1 | ||||
AK316114.1 | ||||
AL121740.1 | CAB57330.1 | |||
BC050727.1 | AAH50727.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
