Assay Details
Target Gene Details
Entrez Gene ID: | 140901 |
Gene Name: | serine/threonine kinase 35 |
Gene Aliases: |
CLIK1, STK35L1 |
Location: |
Chr.20:2098035-2148555 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK35 | XM_011529174.2 | 1 | 890 | XP_011527476.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644996 | Chr.20:2096952 - 2099030 on Build GRCh38 | Loss |
|
| esv2667966 | Chr.20:2096804 - 2098917 on Build GRCh38 | Deletion |
|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
| nsv3258 | Chr.20:2091289 - 2124098 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map