Assay Details
Target Gene Details
Entrez Gene ID: | 116835 |
Gene Name: | heat shock protein family A (Hsp70) member 12B |
Gene Aliases: |
C20orf60 |
Location: |
Chr.20:3732670-3753111 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPA12B | NM_001197327.1 | NP_001184256.1 | ||
| NM_001318322.1 | NP_001305251.1 | |||
| NM_052970.4 | NP_443202.3 | |||
| XM_017027635.1 | XP_016883124.1 | |||
| AK056712.1 | BAB71261.1 | |||
| AK297120.1 | ||||
| AK301294.1 | ||||
| BC090857.1 | AAH90857.1 | |||
| BC110881.1 | AAI10882.1 | |||
| BC117284.1 | ||||
| BC126263.1 | ||||
| BC143932.1 | ||||
| CB990660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833899 | Chr.20:3709295 - 3878350 on Build GRCh38 | Loss |
|
| nsv470536 | Chr.20:3693168 - 3747379 on Build GRCh38 | Loss |
|
| esv3575355 | Chr.20:3667953 - 3752840 on Build GRCh38 | Gain |
|
| nsv585285 | Chr.20:3705789 - 3791211 on Build GRCh38 | Loss |
|
| nsv520565 | Chr.20:3652593 - 3740054 on Build GRCh38 | Loss |
|
| esv3645047 | Chr.20:3734083 - 3737371 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs150590139] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map