Assay Details
Target Gene Details
Entrez Gene ID: | 100528031 |
Gene Name: | FKBP1A-SDCBP2 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.20:1309909-1393172 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FKBP1A-SDCBP2 | NR_037661.1 | |||
| AK309026.1 | ||||
| BM925547.1 |
Target Gene Details
Entrez Gene ID: | 27111 |
Gene Name: | syndecan binding protein 2 |
Gene Aliases: |
SITAC, SITAC18, ST-2 |
Location: |
Chr.20:1309909-1329235 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDCBP2 | NM_001199784.1 | NP_001186713.1 | ||
| NM_080489.4 | NP_536737.3 | |||
| AF159228.1 | AAF80369.1 | |||
| AK296577.1 | ||||
| BC002727.2 | ||||
| BP261525.1 | ||||
| CB147569.1 | ||||
| DB018804.1 |
Target Gene Details
Entrez Gene ID: | 100507495 |
Gene Name: | SDCBP2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.20:1325343-1378735 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDCBP2-AS1 | NR_040047.1 | |||
| NR_040048.1 | ||||
| NR_040049.1 | ||||
| AK055993.1 | ||||
| DA094549.1 | ||||
| DA274215.1 | ||||
| DB454162.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057097 | Chr.20:1050297 - 1333621 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map