Assay Details
Target Gene Details
Entrez Gene ID: | 51643 |
Gene Name: | transmembrane BAX inhibitor motif containing 4 |
Gene Aliases: |
CGI-119, GAAP, LFG4, S1R, ZPRO |
Location: |
Chr.12:66136936-66170072 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMBIM4 | NM_001282606.1 | NP_001269535.1 | ||
| NM_001282609.1 | NP_001269538.1 | |||
| NM_001282610.1 | NP_001269539.1 | |||
| NM_016056.3 | NP_057140.2 | |||
| AF113127.1 | AAF14868.1 | |||
| AF151877.1 | AAD34114.1 | |||
| AF161526.1 | AAF29141.1 | |||
| AF182041.1 | AAG16898.1 | |||
| AF182414.1 | AAG14950.1 | |||
| AK075069.1 | BAC11384.1 | |||
| AK127285.1 | 1 | 749 | ||
| AK225158.1 | ||||
| AL117550.1 | ||||
| BC004401.1 | AAH04401.1 | |||
| BG534013.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629796 | Chr.12:66130674 - 66161690 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map