Assay Details
Target Gene Details
Entrez Gene ID: | 3294 |
Gene Name: | hydroxysteroid 17-beta dehydrogenase 2 |
Gene Aliases: |
EDH17B2, HSD17, SDR9C2 |
Location: |
Chr.16:82035237-82098534 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HSD17B2 | NM_002153.2 | NP_002144.1 | ||
AK223001.1 | BAD96721.1 | |||
AK313109.1 | ||||
AU139034.1 | ||||
BC009581.1 | AAH09581.1 | |||
BC059170.1 | AAH59170.1 | |||
BP270993.1 | ||||
L11708.1 | AAA03562.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
