Assay Details
Target Gene Details
Entrez Gene ID: | 650655 |
Gene Name: | ATP binding cassette subfamily A member 17, pseudogene |
Gene Aliases: |
ABCA17 |
Location: |
Chr.16:2340922-2426699 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCA17P | NR_003574.1 | 1 | 386 | |
| AK095200.1 | 1 | 337 | ||
| DQ266102.1 | 1 | 386 | ||
| DQ660314.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422427 | Chr.16:10001 - 2723164 on Build GRCh38 | Duplication |
|
| nsv571201 | Chr.16:2255538 - 2365929 on Build GRCh38 | Loss |
|
| nsv1131103 | Chr.16:2340199 - 2343199 on Build GRCh38 | Deletion |
|
| nsv827512 | Chr.16:2339663 - 2342655 on Build GRCh38 | Gain |
|
| nsv1052260 | Chr.16:2074546 - 2629655 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75759687] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map