Assay Details
Target Gene Details
Entrez Gene ID: | 2932 |
Gene Name: | glycogen synthase kinase 3 beta |
Gene Aliases: |
- |
Location: |
Chr.3:119821321-120094417 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSK3B | NM_001146156.1 | NP_001139628.1 | ||
NM_002093.3 | NP_002084.2 | |||
XM_006713610.2 | XP_006713673.1 | |||
XM_006713611.2 | XP_006713674.1 | |||
AB451356.1 | ||||
AK290897.1 | ||||
AY123976.1 | AAM88578.1 | |||
BC000251.1 | AAH00251.1 | |||
BC012760.2 | AAH12760.1 | |||
CR536510.1 | CAG38748.1 | |||
EU302497.1 | ||||
L33801.1 | AAA66475.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
