Assay Details
Target Gene Details
Entrez Gene ID: | 2534 |
Gene Name: | FYN proto-oncogene, Src family tyrosine kinase |
Gene Aliases: |
SLK, SYN, p59-FYN |
Location: |
Chr.6:111660332-111873452 on Build GRCh38 |
Assay Gene Location: | Within Intron 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FYN | NM_002037.5 | NP_002028.1 | ||
| NM_153047.3 | NP_694592.1 | |||
| NM_153048.3 | NP_694593.1 | |||
| XM_005266890.3 | XP_005266947.1 | |||
| XM_005266892.3 | XP_005266949.1 | |||
| XM_017010650.1 | XP_016866139.1 | |||
| XM_017010651.1 | XP_016866140.1 | |||
| XM_017010652.1 | XP_016866141.1 | |||
| XM_017010653.1 | XP_016866142.1 | |||
| XM_017010654.1 | XP_016866143.1 | |||
| AB451293.1 | ||||
| AB451426.1 | ||||
| AI187118.1 | ||||
| AK056699.1 | ||||
| BC032496.1 | AAH32496.1 | |||
| M14333.1 | AAC08285.1 | |||
| M14676.1 | AAA36615.1 | |||
| S74774.1 | AAB33113.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5444 | Chr.6:111658463 - 111690809 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map