Assay Details
Target Gene Details
Entrez Gene ID: | 5305 |
Gene Name: | phosphatidylinositol-5-phosphate 4-kinase type 2 alpha |
Gene Aliases: |
PI5P4KA, PIP5K2A, PIP5KII-alpha, PIP5KIIA, PIPK |
Location: |
Chr.10:22534837-22714574 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PIP4K2A | NM_005028.4 | NP_005019.2 | ||
XM_006717450.2 | XP_006717513.1 | |||
XM_011519496.1 | XP_011517798.1 | |||
XM_017016330.1 | XP_016871819.1 | |||
XM_017016331.1 | XP_016871820.1 | |||
XM_017016332.1 | XP_016871821.1 | |||
AK294817.1 | ||||
AK294867.1 | ||||
BC018034.1 | AAH18034.1 | |||
S78798.1 | AAB35041.1 | |||
U14957.2 | AAA64835.2 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
