Assay Details
Target Gene Details
Entrez Gene ID: | 105 |
Gene Name: | adenosine deaminase, RNA specific B2 (inactive) |
Gene Aliases: |
ADAR3, RED2 |
Location: |
Chr.10:1177313-1737476 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADARB2 | NM_018702.3 | NP_061172.1 | ||
AF034837.1 | AAF78094.1 | |||
AI695657.1 | ||||
BC137477.1 | ||||
BC139725.1 | ||||
BC140852.1 |
Target Gene Details
Entrez Gene ID: | 642394 |
Gene Name: | ADARB2 antisense RNA 1 |
Gene Aliases: |
C10orf109, NCRNA00168, bA466B20.1 |
Location: |
Chr.10:1526630-1556984 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADARB2-AS1 | NR_033387.1 | |||
AF086274.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv925n54 | Chr.10:1198080 - 1862404 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv1046041 | Chr.10:1149274 - 2146961 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1041498 | Chr.10:1071260 - 1717996 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv1054555 | Chr.10:1515950 - 1922951 on Build GRCh38 | Gain |
![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
