Assay Details
Target Gene Details
Entrez Gene ID: | 345 |
Gene Name: | apolipoprotein C3 |
Gene Aliases: |
APOCIII, HALP2 |
Location: |
Chr.11:116829908-116833071 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APOC3 | NM_000040.1 | NP_000031.1 | ||
| BC027977.1 | AAH27977.1 | |||
| BC121081.1 | ||||
| BC134419.1 | ||||
| M28613.1 | AAA51760.1 | |||
| M28614.1 | AAA51761.1 | |||
| X00567.1 | CAA25233.1 | |||
| X01388.1 | CAA25644.1 |
More Information
Set Membership: |
|

Genomic Map