Assay Details
Target Gene Details
Entrez Gene ID: | 101929089 |
Gene Name: | uncharacterized LOC101929089 |
Gene Aliases: |
- |
Location: |
Chr.11:118511918-118531094 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101929089 | NR_120572.1 | |||
NR_120573.1 | ||||
NR_120574.1 | ||||
NR_120575.1 | ||||
NR_120576.1 | ||||
AI885123.1 | ||||
AW449653.1 | ||||
AW450539.1 | ||||
AW611756.1 | ||||
BE671460.1 | ||||
BM930081.1 | ||||
BU680885.1 |
Target Gene Details
Entrez Gene ID: | 143941 |
Gene Name: | tetratricopeptide repeat domain 36 |
Gene Aliases: |
HBP21 |
Location: |
Chr.11:118527495-118531025 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TTC36 | NM_001080441.1 | 2 | 199 | NP_001073910.1 |
BC146916.1 | 2 | 222 | ||
BC146921.1 | 2 | 222 | ||
BC171906.1 | 3 | 334 | ||
BC171913.1 | 3 | 357 | ||
EU489483.1 | 2 | 261 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1044281 | Chr.11:118503814 - 118554704 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3627872 | Chr.11:118502100 - 118536457 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
