Assay Details
Target Gene Details
Entrez Gene ID: | 644093 |
Gene Name: | uncharacterized LOC644093 |
Gene Aliases: |
- |
Location: |
Chr.2:47527537-47529164 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCG2040054 | NR_120602.1 | |||
| AI032330.1 | ||||
| AI150334.1 |
Target Gene Details
Entrez Gene ID: | 56660 |
Gene Name: | potassium two pore domain channel subfamily K member 12 |
Gene Aliases: |
K2p12.1, THIK-2, THIK2 |
Location: |
Chr.2:47520775-47570331 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK12 | NM_022055.1 | NP_071338.1 | ||
| AF287302.1 | AAG32313.1 | |||
| BC047749.1 | AAH47749.1 |
Target Gene Details
Entrez Gene ID: | 4436 |
Gene Name: | mutS homolog 2 |
Gene Aliases: |
COCA1, FCC1, HNPCC, HNPCC1, LCFS2 |
Location: |
Chr.2:47403067-47634501 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MSH2 | XM_005264332.3 | XP_005264389.2 | ||
| XM_011532867.1 | XP_011531169.1 | |||
| AK223284.1 | BAD97004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999576 | Chr.2:47278977 - 47546821 on Build GRCh38 | Gain |
|
| nsv998314 | Chr.2:47493281 - 47732029 on Build GRCh38 | Loss |
|
| nsv1004828 | Chr.2:46829710 - 47587343 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map