Assay Details
Target Gene Details
Entrez Gene ID: | 60509 |
Gene Name: | ATP/GTP binding protein like 5 |
Gene Aliases: |
CCP5, RP75 |
Location: |
Chr.2:27051423-27070622 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AGBL5 | NM_021831.5 | 15 | 3167 | NP_068603.4 |
| NR_104246.1 | 16 | 3214 | ||
| XM_005264477.2 | 14 | 3033 | XP_005264534.1 | |
| AI990522.1 | 1 | 71 | ||
| AL833844.1 | 15 | 3109 | CAD38704.1 | |
| BC007415.2 | 16 | 3156 |
Target Gene Details
Entrez Gene ID: | 100128731 |
Gene Name: | oligosaccharyltransferase complex subunit 4, non-catalytic |
Gene Aliases: |
- |
Location: |
Chr.2:27070472-27071699 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OST4 | NM_001134693.1 | 3 | 407 | NP_001128165.1 |
| AA772256.1 | 1 | 103 | ||
| AL049447.1 | 3 | 333 | ||
| BF697310.1 | 3 | 407 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv6744n54 | Chr.2:26973431 - 27151141 on Build GRCh38 | Loss |
|
| nsv817673 | Chr.2:27037601 - 27163931 on Build GRCh38 | Gain |
|
| nsv833603 | Chr.2:27059106 - 27263631 on Build GRCh38 | Loss |
|
| nsv455974 | Chr.2:26989775 - 27128411 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map