Assay Details
Target Gene Details
Entrez Gene ID: | 8837 |
Gene Name: | CASP8 and FADD like apoptosis regulator |
Gene Aliases: |
CASH, CASP8AP1, CLARP, Casper, FLAME, FLAME-1, FLAME1, FLIP, I-FLICE, MRIT, c-FLIP, c-FLIPL, c-FLIPR, c-FLIPS |
Location: |
Chr.2:201116015-201172688 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFLAR | NM_001127184.2 | NP_001120656.1 | ||
| NM_001202516.1 | NP_001189445.1 | |||
| NM_001202517.1 | NP_001189446.1 | |||
| NM_001202519.1 | NP_001189448.1 | |||
| NM_001308043.1 | NP_001294972.1 | |||
| NM_003879.5 | NP_003870.4 | |||
| XM_017005189.1 | XP_016860678.1 | |||
| XM_017005192.1 | XP_016860681.1 | |||
| XM_017005193.1 | 1 | 528 | XP_016860682.1 | |
| XM_017005195.1 | XP_016860684.1 | |||
| XM_017005196.1 | XP_016860685.1 | |||
| XM_017005197.1 | 1 | 528 | XP_016860686.1 | |
| AB209600.1 | BAD92837.1 | |||
| AF005774.1 | AAC15825.1 | |||
| AF009616.1 | AAB70909.1 | |||
| AF010127.1 | AAB64110.1 | |||
| AK296036.1 | ||||
| AK297387.1 | ||||
| AK315924.1 | ||||
| BC001602.1 | AAH01602.1 | |||
| BX457155.2 | ||||
| DC375144.1 | ||||
| U97074.1 | AAC51622.1 | |||
| Y14039.1 | CAA74366.1 | |||
| Y14040.1 | CAA74367.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv954818 | Chr.2:201115378 - 201131577 on Build GRCh38 | Duplication |
|
| nsv584160 | Chr.2:201039216 - 201298760 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map