Assay Details
Target Gene Details
Entrez Gene ID: | 23380 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 2 |
Gene Aliases: |
ARHGAP34, FNBP2, SRGAP2A, SRGAP3 |
Location: |
Chr.1:206203464-206464443 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRGAP2 | NM_001170637.3 | NP_001164108.1 | ||
| NM_001300952.1 | NP_001287881.1 | |||
| NM_015326.4 | NP_056141.2 | |||
| XM_005277510.2 | XP_005277567.1 | |||
| XM_005277511.2 | XP_005277568.1 | |||
| XM_005277514.1 | XP_005277571.1 | |||
| XM_005277515.1 | XP_005277572.1 | |||
| XM_011509354.1 | XP_011507656.1 | |||
| XM_011509355.1 | XP_011507657.1 | |||
| XM_011509356.2 | XP_011507658.1 | |||
| XM_011509359.1 | XP_011507661.1 | |||
| XM_017000838.1 | XP_016856327.1 | |||
| XM_017000840.1 | XP_016856329.1 | |||
| XM_017000841.1 | XP_016856330.1 | |||
| AB007925.1 | ||||
| AK295845.1 | ||||
| BC132872.1 | ||||
| BC132874.1 | ||||
| BC144343.1 |
Target Gene Details
Entrez Gene ID: | 647135 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 2B |
Gene Aliases: |
SRGAP2L, SRGAP2P2 |
Location: |
Chr.1:144887191-145093441 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRGAP2B | NM_001271870.2 | NP_001258799.2 | ||
| XM_005277421.3 | XP_005277478.1 | |||
| XM_017002085.1 | XP_016857574.1 | |||
| XM_017002086.1 | XP_016857575.1 | |||
| XM_017002087.1 | XP_016857576.1 | |||
| XM_017002088.1 | XP_016857577.1 | |||
| XM_017002089.1 | XP_016857578.1 | |||
| XM_017002090.1 | XP_016857579.1 | |||
| XM_017002091.1 | XP_016857580.1 | |||
| XM_017002092.1 | XP_016857581.1 |
Target Gene Details
Entrez Gene ID: | 653464 |
Gene Name: | SLIT-ROBO Rho GTPase activating protein 2C |
Gene Aliases: |
SRGAP2P1 |
Location: |
Chr.1:121184967-121392819 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRGAP2C | NM_001271872.1 | NP_001258801.1 | ||
| XM_005277497.3 | XP_005277554.1 | |||
| XM_017002149.1 | XP_016857638.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761652 | Chr.1:144928063 - 145023143 on Build GRCh38 | Gain |
|
| nsv1015024 | Chr.1:144995424 - 145023143 on Build GRCh38 | Gain |
|
| dgv312n100 | Chr.1:144950111 - 145023143 on Build GRCh38 | Gain |
|
| dgv40e214 | Chr.1:144915703 - 145023143 on Build GRCh38 | Gain |
|
| nsv547651 | Chr.1:144946566 - 145023143 on Build GRCh38 | Loss |
|
| nsv1118446 | Chr.1:144971906 - 145009301 on Build GRCh38 | Duplication |
|
| nsv1010171 | Chr.1:144979963 - 145023143 on Build GRCh38 | Gain |
|
| nsv946222 | Chr.1:144914153 - 145023143 on Build GRCh38 | Duplication |
|
| nsv1003232 | Chr.1:144970891 - 145023143 on Build GRCh38 | Gain+Loss |
|
| nsv1003402 | Chr.1:144963817 - 145023143 on Build GRCh38 | Loss |
|
| esv2516798 | Chr.1:144984771 - 145022677 on Build GRCh38 | Gain |
|
| nsv1010181 | Chr.1:144942033 - 145023143 on Build GRCh38 | Gain+Loss |
|
More Information
Additional Information:
This assay has 3 genomic targets in NCBI build GRCh38
Set Membership: |
|

Genomic Map