Assay Details
Target Gene Details
Entrez Gene ID: | 56342 |
Gene Name: | peter pan homolog (Drosophila) |
Gene Aliases: |
BXDC3, SSF, SSF-1, SSF1, SSF2 |
Location: |
Chr.19:10106223-10111299 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPAN | NM_020230.5 | NP_064615.3 | ||
| AJ292529.1 | CAB99252.1 | |||
| BC009833.2 | AAH09833.1 | |||
| BC033202.1 | AAH33202.1 |
Target Gene Details
Entrez Gene ID: | 692312 |
Gene Name: | PPAN-P2RY11 readthrough |
Gene Aliases: |
- |
Location: |
Chr.19:10106223-10115389 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPAN-P2RY11 | NM_001040664.2 | NP_001035754.1 | ||
| NM_001198690.1 | NP_001185619.1 | |||
| AJ300588.1 | CAC18877.1 |
Target Gene Details
Entrez Gene ID: | 100113382 |
Gene Name: | small nucleolar RNA, C/D box 105B |
Gene Aliases: |
- |
Location: |
Chr.19:10109749-10109840 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SNORD105B | NR_003688.1 | 1 | 49 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833743 | Chr.19:10077649 - 10251913 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map