Assay Details
Target Gene Details
Entrez Gene ID: | 5789 |
Gene Name: | protein tyrosine phosphatase, receptor type D |
Gene Aliases: |
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA |
Location: |
Chr.9:8314246-10612840 on Build GRCh38 |
Assay Gene Location: | Within Intron 11 |
Target Gene Details
Entrez Gene ID: | 101929407 |
Gene Name: | PTPRD antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:8858018-8861727 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTPRD-AS1 | NR_121599.1 | |||
| NR_121600.1 | ||||
| AK094342.1 | 2 | 599 | ||
| DA788948.1 | ||||
| DB495850.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015898 | Chr.9:8753534 - 9011443 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map