Assay Details
Target Gene Details
Entrez Gene ID: | 3569 |
Gene Name: | interleukin 6 |
Gene Aliases: |
BSF-2, BSF2, CDF, HGF, HSF, IFN-beta-2, IFNB2, IL-6 |
Location: |
Chr.7:22725889-22732002 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL6 | NM_000600.4 | NP_000591.1 | ||
| NM_001318095.1 | NP_001305024.1 | |||
| XM_005249745.4 | XP_005249802.1 | |||
| XM_011515390.2 | XP_011513692.1 | |||
| A09363.1 | CAA00839.1 | |||
| AK298013.1 | ||||
| AK298077.1 | ||||
| AK301141.1 | ||||
| BC015511.1 | AAH15511.1 | |||
| BT019748.1 | AAV38553.1 | |||
| BT019749.1 | AAV38554.1 | |||
| CD013918.1 | ||||
| CR450296.1 | CAG29292.1 | |||
| HY076725.1 | ||||
| M14584.1 | AAA52728.1 | |||
| M18403.1 | AAA52729.1 | |||
| M29150.1 | AAA59154.1 | |||
| M54894.1 | AAC41704.1 | |||
| S56892.1 | AAD13886.1 | |||
| X04403.1 | CAA27991.1 | |||
| X04430.1 | CAA28026.1 | |||
| X04602.1 | CAA28268.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759518 | Chr.7:22715965 - 22881291 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map