Assay Details
Target Gene Details
| Entrez Gene ID: | 1124 | 
| Gene Name: | chimerin 2 | 
| Gene Aliases: | ARHGAP3, BCH, CHN2-3, RHOGAP3 | 
| Location: | Chr.7:29146547-29514335 on Build GRCh38 | 
| Assay Gene Location: | Overlaps Intron 1 - Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| CHN2 | NM_001293069.1 | NP_001279998.1 | ||
| XM_011515105.2 | XP_011513407.1 | |||
| XM_011515106.2 | XP_011513408.1 | |||
| XM_017011721.1 | XP_016867210.1 | |||
| AK294271.1 | ||||
| DA190643.1 | ||||
| GQ924106.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3612685 | Chr.7:28997136 - 29209454 on Build GRCh38 | Loss |  LOC100506497  CPVL  CHN2  LOC102724484  LOC101928168 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  DGV Variation | 
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top