Assay Details
Target Gene Details
Entrez Gene ID: | 64951 |
Gene Name: | mitochondrial ribosomal protein S24 |
Gene Aliases: |
HSPC335, MRP-S24, S24mt, bMRP-47, bMRP47 |
Location: |
Chr.7:43866558-43869546 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS24 | NM_032014.2 | NP_114403.1 | ||
| AF161453.1 | AAF29013.1 | |||
| AK054982.1 | ||||
| BC012167.1 | AAH12167.1 | |||
| BC054865.1 | AAH54865.1 | |||
| CA413186.1 |
Target Gene Details
Entrez Gene ID: | 100534592 |
Gene Name: | URGCP-MRPS24 readthrough |
Gene Aliases: |
- |
Location: |
Chr.7:43866558-43906632 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| URGCP-MRPS24 | NM_001204871.1 | NP_001191800.1 | ||
| AI027170.1 | ||||
| FY210844.1 | ||||
| FY212401.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428165 | Chr.7:43855828 - 44172960 on Build GRCh38 | Gain |
|
| nsv830980 | Chr.7:43865998 - 44078539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map