Assay Details
Target Gene Details
Entrez Gene ID: | 8848 |
Gene Name: | TSC22 domain family member 1 |
Gene Aliases: |
Ptg-2, TGFB1I4, TSC22 |
Location: |
Chr.13:44432143-44576565 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TSC22D1 | NM_001243798.1 | NP_001230727.1 | ||
| NM_001243799.1 | NP_001230728.1 | |||
| NM_183422.3 | NP_904358.2 | |||
| AB082525.1 | ||||
| AK027071.1 | ||||
| AK091854.1 | ||||
| BC136475.1 | ||||
| BC136482.1 | ||||
| BI463370.1 | ||||
| CR627459.1 | CAH10539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3631934 | Chr.13:44468293 - 44468970 on Build GRCh38 | Loss |
|
| esv2677067 | Chr.13:44468296 - 44468969 on Build GRCh38 | Deletion |
|
| esv23675 | Chr.13:44468426 - 44468984 on Build GRCh38 | Loss |
|
| nsv958104 | Chr.13:44468315 - 44469077 on Build GRCh38 | Deletion |
|
| nsv561575 | Chr.13:44468473 - 44468929 on Build GRCh38 | Loss |
|
| esv2747332 | Chr.13:44468309 - 44468974 on Build GRCh38 | Deletion |
|
| esv3550396 | Chr.13:44468252 - 44469032 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map