Assay Details
Target Gene Details
Entrez Gene ID: | 5146 |
Gene Name: | phosphodiesterase 6C |
Gene Aliases: |
ACHM5, COD4, PDEA2 |
Location: |
Chr.10:93612588-93665673 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDE6C | NM_006204.3 | NP_006195.3 | ||
| XM_017016322.1 | XP_016871811.1 | |||
| U31973.1 | AAA96392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv21639 | Chr.10:93628988 - 93630298 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map