Assay Details
Target Gene Details
Entrez Gene ID: | 26000 |
Gene Name: | TBC1 domain family member 10B |
Gene Aliases: |
EPI64B, FP2461 |
Location: |
Chr.16:30357101-30370258 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TBC1D10B | NM_015527.3 | 1 | 126 | NP_056342.3 |
| XM_011545789.1 | 1 | 183 | XP_011544091.1 | |
| XM_011545790.2 | 1 | 183 | XP_011544092.1 | |
| AI672923.1 | 1 | 189 | ||
| BY795947.2 | 1 | 103 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26390 | Chr.16:30369673 - 30370512 on Build GRCh38 | Loss |
|
| nsv827612 | Chr.16:30360217 - 30418541 on Build GRCh38 | Gain |
|
| esv2758642 | Chr.16:30062634 - 30371959 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map