Assay Details
Target Gene Details
Entrez Gene ID: | 245 |
Gene Name: | arachidonate 12-lipoxygenase pseudogene 2 |
Gene Aliases: |
ALOX12E |
Location: |
Chr.17:6853576-6900349 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALOX12P2 | NR_002710.2 | 7 | 1314 | |
| NR_120453.1 | 5 | 674 | ||
| AF020774.1 | 5 | 675 | ||
| AL832768.1 | 7 | 1031 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv21524 | Chr.17:6895300 - 6895832 on Build GRCh38 | Loss |
|
| esv2761944 | Chr.17:6872184 - 6952241 on Build GRCh38 | Loss |
|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv1060335 | Chr.17:6845771 - 6960016 on Build GRCh38 | Loss |
|
| dgv3095n100 | Chr.17:6872614 - 6962694 on Build GRCh38 | Loss |
|
| nsv521515 | Chr.17:6855195 - 6957288 on Build GRCh38 | Loss |
|
| nsv518777 | Chr.17:6895059 - 6898945 on Build GRCh38 | Gain |
|
| esv3582504 | Chr.17:6846017 - 6962678 on Build GRCh38 | Loss |
|

Genomic Map