Assay Details
Target Gene Details
Entrez Gene ID: | 51088 |
Gene Name: | kelch like family member 5 |
Gene Aliases: |
- |
Location: |
Chr.4:39044796-39143101 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL5 | NM_001007075.2 | NP_001007076.1 | ||
| NM_001171654.1 | NP_001165125.1 | |||
| XM_005262655.4 | XP_005262712.1 | |||
| XM_011513699.2 | XP_011512001.1 | |||
| XM_011513700.2 | XP_011512002.1 | |||
| XM_011513701.2 | XP_011512003.1 | |||
| XM_017008273.1 | XP_016863762.1 | |||
| XM_017008274.1 | XP_016863763.1 | |||
| XM_017008275.1 | XP_016863764.1 | |||
| XM_017008276.1 | XP_016863765.1 | |||
| AB208820.1 | BAD92057.1 | |||
| BC053860.1 | AAH53860.1 | |||
| DB451501.1 | ||||
| DR006899.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1073752 | Chr.4:39044379 - 39045980 on Build GRCh38 | Deletion |
|
| nsv594013 | Chr.4:39044710 - 39045265 on Build GRCh38 | Gain+Loss |
|
| esv28537 | Chr.4:39044683 - 39045398 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map