Assay Details
Target Gene Details
Entrez Gene ID: | 110948 |
Gene Name: | holocarboxylase synthetase (biotin- [propriony-Coenzyme A-carboxylase (ATP-hydrolysing)] ligase) |
Gene Aliases: |
410I21.SP6, D16Jhu34 |
Location: |
Chr.16:94129195-94313905 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hlcs | NM_139145.4 | 5 | 1470 | NP_631884.1 |
| AB066227.1 | 4 | 1330 | BAB68213.1 | |
| AK052424.1 | 6 | 1766 | ||
| AK158212.1 | 6 | 1587 | BAE34407.1 | |
| AK164351.1 | 6 | 1751 | ||
| AK168925.1 | 2 | 365 | BAE40738.1 | |
| BC050090.1 | 5 | 1471 | AAH50090.1 |
More Information
Set Membership: |
|

Genomic Map