Assay Details
Target Gene Details
Entrez Gene ID: | 20658 |
Gene Name: | Son DNA binding protein |
Gene Aliases: |
2900011L12Rik, AA409051, AU067731, C81487, mKIAA1019, nrebp |
Location: |
Chr.16:91647824-91679195 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Son | NM_019973.2 | 3 | 1076 | NP_064357.2 |
| NM_178880.4 | 3 | 1076 | NP_849211.3 | |
| AB546195.1 | 3 | 1011 | ||
| AF193607.1 | 3 | 1020 | AAF23121.1 | |
| BC046419.1 | 3 | 1015 | AAH46419.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map