Assay Details
Target Gene Details
Entrez Gene ID: | 23872 |
Gene Name: | E26 avian leukemia oncogene 2, 3' domain |
Gene Aliases: |
AU022856, Ets-2 |
Location: |
Chr.16:95702407-95721049 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ets2 | NM_011809.3 | 10 | 3482 | NP_035939.3 |
| AK005402.1 | 1 | 639 | ||
| AK014602.2 | 10 | 3404 | ||
| AK030537.1 | 6 | 2784 | BAC27010.1 | |
| AK038001.1 | 1 | 3243 | ||
| AK038348.1 | 10 | 3469 | BAE20535.1 | |
| AK128933.1 | 11 | 3356 | ||
| AK143687.1 | 10 | 3434 | BAE25498.1 | |
| AK144974.1 | 10 | 3433 | BAE26161.1 | |
| AK145852.1 | 10 | 3318 | BAE26698.1 | |
| AK151315.1 | 10 | 3433 | BAE30296.1 | |
| AK152750.1 | 10 | 3436 | ||
| AK153478.1 | 10 | 3434 | BAE32028.1 | |
| AK204609.1 | 1 | 309 | ||
| BC005486.1 | 10 | 3403 | AAH05486.1 | |
| BC005504.1 | 10 | 3343 | AAH05504.1 | |
| BE134375.1 | 1 | 79 |
More Information
Set Membership: |
|

Genomic Map