Assay Details
Target Gene Details
Entrez Gene ID: | 54720 |
Gene Name: | regulator of calcineurin 1 |
Gene Aliases: |
Adapt78, CALP1L, CSP1, DSC1, Dscr1, MCIP1, RCN1 |
Location: |
Chr.16:92391951-92466169 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rcan1 | NM_001081549.2 | NP_001075018.1 | ||
| NM_019466.4 | NP_062339.2 | |||
| AF237789.1 | AAF63485.1 | |||
| AF237790.1 | AAF63486.1 | |||
| AF260717.1 | AAF70343.1 | |||
| AF263239.1 | AAF72701.1 | |||
| AF263240.1 | AAF72702.1 | |||
| AF282255.1 | AAF91461.1 | |||
| AK077283.1 | BAC36729.1 | |||
| AK146764.1 | BAE27416.1 | |||
| AY325904.1 | AAP96744.1 | |||
| BC013551.1 | AAH13551.1 |
More Information
Set Membership: |
|

Genomic Map