Assay Details
Target Gene Details
Entrez Gene ID: | 93961 |
Gene Name: | UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 5 |
Gene Aliases: |
1190002B21Rik, AU045265, b3Galt-V |
Location: |
Chr.16:96235801-96319859 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3galt5 | NM_001122993.1 | NP_001116465.1 |
More Information
Set Membership: |
|

Genomic Map